Learn More
Amsta Rostfritt Stainless Steel Containers With Lid and 3 Clamps
Click to view available options
Volume (Metric):
1 L
2 L
3 L
5 L
6 L
8 L
10 L
20 L
Unit Size:
Each
Specifications
Specifications
| Product Type | Container with Lid and 3 Clamps |
| Material | Stainless Steel |
| Dimensions (L x W) | 155 x 180 mm |
| Volume (Metric) | 3 L |