Learn More
Grant Instruments Stacking Kit
Product Code. 30274355
Click to view available options
Holds:
2 Rollers
3 Rollers
Unit Size:
Each
Product Code. 30274355
Brand: Grant Instruments CTR SK2
Only null left
Add to basket
Add to basket
Stacking kit for CTR-6.
  • CTR stacking kit
  • Available for stacking 2 or 3 CTR-6's
TRUSTED_SUSTAINABILITY
Specifications
Holds 2 Rollers
Product Type Stacking Kit
Material Stainless Steel
Diameter (English) 0.3 in.
Diameter (Metric) 8 mm
Weight (English) 1.2 lb.
Weight (Metric) 0.5 kg
Color Metal
For Use With (Equipment) CTR-6
Quantity 1 Ea.