missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Stablcal™ Formazine Reference Suspension I 3 NTU
This pre-packaged and ready to use Stablcal Formazine Reference Suspension with a volume of 100 mL and a concentration of 3 NTUs is certified and stable for 2 years.
Brand: Hach Lange™ 2896742
This item is not returnable.
View return policy
Description
- Stabilised Formazine reference suspension for the determination of lucidity/opalescence according to European Pharmacopeia 6.0.
- Pre-packaged and ready to use
- Stable for 2 years
- Certified quality
- Concentration: 3 NTU
- Volume / Package size: 100 mL
Specifications
Stablcal Formazine Reference Suspension I | |
3 NTU | |
100 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- StablCal Stabilized Formazin Turbidity Standard; 3 NTU
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : methenamine; hexamethylenetetramine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction