missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SS Agar (Dehydrated)

Product Code. p-200112234
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10650705

Brand: Thermo Scientific™ CM0099B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and identify Salmonella and Shigella species from clinical or food samples with this selective medium.

Isolate and identify Salmonella and Shigella species from clinical or food samples with Thermo Scientific™ Oxoid™ SS Agar (Dehydrated). Differentiation of Salmonella and Shigella is achieved by incorporation of lactose in the medium. Organisms which ferment lactose produce acid which, in the presence of the neutral red indicator, results in the formation of red colonies. Salmonella, Shigella, and other non-lactose-fermenting organisms appear as transparent or straw coloured colonies on SS Agar.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.

SS Agar is a differential, selective medium for the isolation of Shigella and Salmonella species from pathological specimens, suspected foodstuffs etc.

  • Easy-to-read: lactose fermenters (red colonies) are easy to differentiate from non-lactose fermenters like Shigella and Salmonella (straw colorless colonies).
  • Selective: due to the inclusion of selective inhibitory substances brilliant green, bile salts, thiosulphate, and citrate, most Gram-positive and coliform organisms are inhibited.

Thiosuplate in combination with iron acts as an indicator of sulphide production, which is indicated by blackening in the centers of the colonies.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description SS Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 7.9L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.