Learn More
Buerkle™ Squeeze-Fix Hose Clamp
Shop All Bürkle Products
Click to view available options
Tubing Size:
1 to 5 mm
3 to 10 mm
6 to 15 mm
Overall Length (Metric):
25 mm
40 mm
55 mm
Unit Size:
Each
Description
Using these clips, hoses can be compressed partially or completely as required.
- Can be operated with one hand
- Made from polyoxymethylene plastic (POM)
- Does not rust
- Permanently elastic and hygienic
- White
- Sterilizable
Specifications
Specifications
| Material | POM |
| Overall Length (Metric) | 40 mm |
| For Use With | Hoses |
| Tubing Size | 3 to 10 mm |
| Color | White |
| Product Type | Hose Clamp |