Learn More
Julabo™ Spring Clamps for Shaking Water Bath
Click to view available options
Type:
Spring Clamp
Spring Clamp Set
Holds:
10 mL Erlenmeyer flask
100 mL Erlenmeyer flask
1000 mL Erlenmeyer flask
12-135 mm and 5-190mm L springs
200 to 250 mL Erlenmeyer flasks
25 mL Erlenmeyer flask
300 mL Erlenmeyer flask
50 mL Erlenmeyer flask
500 mL Erlenmeyer flask
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | SW22, SW23 shaking water bath |
| Type | Spring Clamp |
| Holds | 25 mL Erlenmeyer flask |