Learn More
Edmund Bühler GmbH™ Spring Clamp
Product Code. p-8013055
Click to view available options
Holds:
10 mL Flask
500 mL
Unit Size:
Each
Product Code. 10049390
Brand: Edmund Bühler GmbH™ 0009 647
Only null left
Add to basket
Add to basket
Fasten Erlenmeyer flasks on the universal tray.
  • Stainless steel construction
  • For fastening Erlenmeyer flasks on the universal tray
  • The sizes are related to Erlenmeyer flasks, but are also suitable for round-bottom flasks, beakers, etc
Specifications
Holds 500 mL
Product Type Spring Clamp
Material Stainless Steel