Learn More
Edmund Bühler GmbH™ Spring Clamp
Product Code. 10596391
Click to view available options
Holds:
10 mL Flask
500 mL
Unit Size:
Each
Product Code. 10596391
Brand: Edmund Bühler GmbH™ 0009 642
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fasten Erlenmeyer flasks on the universal tray.
  • Stainless steel construction
  • For fastening Erlenmeyer flasks on the universal tray
  • The sizes are related to Erlenmeyer flasks, but are also suitable for round-bottom flasks, beakers, etc
Specifications
Holds 10 mL Flask
Product Type Spring Clamp