Learn More
Thermo Scientific Chemicals Spironolactone, 99%
€ 66.60 - € 224.00
Chemical Identifiers
CAS | 52-01-7 |
---|---|
Molecular Formula | C24H32O4S |
Molecular Weight (g/mol) | 416.57 |
MDL Number | MFCD00082250 |
InChI Key | LXMSZDCAJNLERA-ZHYRCANASA-N |
Synonym | spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren |
PubChem CID | 5833 |
ChEBI | CHEBI:9241 |
IUPAC Name | S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate |
SMILES | CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10082800
|
Thermo Scientific Chemicals
207460010 |
1 g | Glass bottle |
€ 66.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10424165
|
Thermo Scientific Chemicals
207460050 |
5 g | Glass bottle |
€ 224.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52-01-7 | |
416.57 | |
LXMSZDCAJNLERA-ZHYRCANASA-N | |
5833 | |
S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate |
C24H32O4S | |
MFCD00082250 | |
spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren | |
CHEBI:9241 | |
CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C |
Specifications
52-01-7 | |
White to White-Yellow | |
Authentic | |
C24H32O4S | |
15, 8887 | |
− 37.00 | |
Solubility in water: practically insoluble. Other solubilities: soluble in most common organic solvents | |
CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C | |
416.57 | |
CHEBI:9241 | |
99% | |
Spironolactone, 99% |
198.0°C to 207.0°C | |
0.5% max. (105°C, 2 hrs) | |
98.5% min. (HPLC) | |
MFCD00082250 | |
−37° (20°C c=1,CHCl3) | |
spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren | |
LXMSZDCAJNLERA-ZHYRCANASA-N | |
S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate | |
5833 | |
416.57 | |
Powder |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
May damage fertility.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-133-6
missing translation for 'rtecsNumber' : TU4725000
missing translation for 'tsca' : TSCA
- Spironolactone
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1B
Hazard Statement
- H360F-May damage fertility.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only