Learn More
VACUUBRAND™ Special Oil
Brand: VACUUBRAND™ 20687700
65.37 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Specifications
Special Oil TWII | |
Bottle | |
1 Ea. | |
1 L |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
VACUUBRAND™ Special Oil > Volume: 1L