missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten & Graf GmbH Spare Valve Block Unit for FORTUNA™ POLYFIX™ Dispenser
Poulten & Graf GmbH Spare Valve Block Unit for FORTUNA™ POLYFIX™ Dispenser comes with dispensing cylinder and discharge tube.
€ 99.15 - € 114.00
Specifications
Quantity | 1 |
---|---|
Type | Valve Block Unit for POLYFIX Dispenser |
For Use With (Equipment) | POLYFIX dispenser |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10004450
|
Poulten & Graf GmbH
101 07538 |
2 to 10 mL |
€ 99.15
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
10196420
|
Poulten & Graf GmbH
101 07548 |
10 to 50 mL |
€ 114.00
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
Description
- Complete with dispensing cylinder
- Includes discharge tube
Specifications
1 | |
Valve Block Unit for POLYFIX Dispenser | |
POLYFIX dispenser |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title