Learn More
Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths
Provides for intense yet gentle cleaning of technical parts. Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths feature tanks and drip-proof housings of stainless steel.
Brand:  Bandelin Electronic™ 322
 View more versions of this product
Product Code. 10776871
  • € 3125.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Description
Provides for intense yet gentle cleaning of technical parts. Bandelin Electronic™ Sonorex™ Super RK Series Ultrasonic Baths feature tanks and drip-proof housings of stainless steel.
  • SweepTec™ for permanent sound field oscillation
  • LCT-transducers - working frequency of 36 kHz
  • All units are CE-marked
  • Degree of protection IP 32
  • Timer for 1-15 minutes or continuous operation
  • Excellent for technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Specifications
1.4 A
230 V
500 x 300 x 200 mm (LxWxD)
RK 1028
Stainless Steel
28 L
For cleaning of technical glassware, test sieves, medical instruments, metal parts, electronic components, degassing, emulsifying, sample preparation, reaction acceleration