Learn More
Sonoco™ ThermoSafe InsulatedShipper-PUR, Packed in Pallet
Product Code. 17818703
Click to view available options
Dimensions (L x W x H):
19 x 14.75 x 22.25 in.
22.75 x 19.5 x 19.5 in.
23.25 x 23 x 24 in.
Unit Size:
Pack of 12
Pack of 18
Pack of 8
Product Code. 17818703
Brand: Sonoco™ E126
This item requires a price update prior to being ordered.
Request a Price for this item here.
High performance polyurethane containers meet the needs of the most demanding temperature sensitive payloads
Polyurethane insulation offers greater insulating power than equivalent thicknesses of EPS containers. Thick walls and 200 lb. outer corrugate offer protective shock absorption and cushioning during shipping and handling.
  • Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
  • Provides extensive protection for long range journeys
  • Durable one-piece closed-cell polyurethane foam insulated shippers are CFC and HCFC-free—comply with international clean air regulations
  • Proprietary process laminates highly efficient insulation core to inner and outer corrugate walls
  • Withstands crush compression in excess of 5000 lb.
  • Flexible polyurethane foam plug seals the top, cushions and insulates contents, vents dry ice
  • R-value is +1.5 times EPS R-value, per inch of thickness
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 56.5 cm
Height (English) 22.25 in.
Length (Metric) 48.2 cm
Length (English) 19 in.
Width (English) 14.75 in.
Dimensions (L x W x H) 19 x 14.75 x 22.25 in.
For Use With (Application) Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
Length (English) Interior 14.25 in.
Length (Metric) Interior 36.1 cm
Width (English) Interior 37.4 cm
Width (Metric) Interior 25.4 cm
Height (English) Interior 15.25 in.
Height (Metric) Interior 38.7 cm