Learn More
Fisherbrand™ Solid Neoprene Rubber Bungs
Product Code. 11527103 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Bottom:
3 mm
4 mm
6 mm
7 mm
9 mm
10 mm
11 mm
13 mm
17 mm
19 mm
25 mm
27 mm
33 mm
39 mm
43 mm
49 mm
53 mm
57 mm
62 mm
67 mm
71 mm
75 mm
79 mm
86 mm
92 mm
99 mm
Diameter (Metric) Top:
4.5 mm
5.8 mm
8 mm
9 mm
11.5 mm
12.5 mm
14 mm
16 mm
20.3 mm
22.5 mm
28.5 mm
31 mm
38.5 mm
44.5 mm
49 mm
55 mm
59.5 mm
64 mm
69 mm
74 mm
78 mm
85 mm
89 mm
96 mm
102 mm
109 mm
Length (Metric):
12 mm
14 mm
16 mm
20 mm
24 mm
26 mm
28 mm
32 mm
38.5 mm
42 mm
45.5 mm
49 mm
54 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
Product Code. 11527103
Only null left
Add to basket
Add to basket
Used for laboratory purposes.
  • Made up of neoprene rubber
  • Hardness is 46/55 IRHD
Specifications
For Use With (Application) For laboratory use
Hardness 46/55 IRHD
Length (Metric) 28 mm
Quantity 10/Pk.
Color Gray
Diameter (Metric) Bottom 25 mm
Diameter (Metric) Top 28.5 mm
Material Neoprene
Product Type Bung