missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sofosbuvir impurity A, MedChemExpress
MedChemExpress Sofosbuvir impurity A, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
€ 1321.65
Specifications
CAS | 1496552-16-9 |
---|---|
Molecular Formula | C22H29FN3O9P |
Molecular Weight (g/mol) | 529.45 |
Description
Sofosbuvir impurity A, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.Specifications
1496552-16-9 | |
C22H29FN3O9P | |
529.45 |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only