missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
€ 109.55 - € 274.37
Chemical Identifiers
CAS | 10049-21-5 |
---|---|
Molecular Formula | H5NaO5P |
Molecular Weight (g/mol) | 139.00 |
MDL Number | MFCD00149208 |
InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
PubChem CID | 516949 |
IUPAC Name | sodium;dihydrogen phosphate;hydrate |
SMILES | O.[Na+].OP(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12625087
|
Thermo Scientific Chemicals
447312500 |
250 g | Plastic bottle |
€ 109.55
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12615087
|
Thermo Scientific Chemicals
447310010 |
1 kg | Plastic bottle |
€ 274.37
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
10049-21-5 | |
139.00 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
516949 | |
O.[Na+].OP(O)(O)=O |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
sodium;dihydrogen phosphate;hydrate |
Specifications
100.0°C | |
99.5% | |
NaH2PO4·H2O | |
15,879 | |
Clear colorless | |
O.[Na+].OP(O)(O)=O | |
139.00 | |
137.99 | |
Biochemical |
10049-21-5 | |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium;dihydrogen phosphate;hydrate | |
516949 | |
99.5% | |
Sodium phosphate, monobasic monohydrate |
Safety and Handling
Product Identifier
- Sodium phosphate, monobasic monohydrate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry