missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrite, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E250, 99 to 100.5% (calc. to the dried substance), Honeywell Fluka™

Product Code. p-7071810 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
50 kg
Unit Size:
1kg
50kg
5kg
This item is not returnable. View return policy

Product Code. 15641330

Brand: Honeywell-Fluka 134471KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E250, 99-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O

Specifications

Quantity 1 kg
Linear Formula NaNO2
UN Number 1500
Formula Weight 69g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Ignition Point 914°F
Chemical Name or Material Sodium Nitrite, Meets analytical specification of Ph. Eur.BP, USP, FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.