missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorodifluoroacetate, 97%, Thermo Scientific Chemicals

Product Code. 11452408
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.

Solubility
Soluble in dimethoxyethane, diglyme (0.60 g mL-1), triglyme, dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1895-39-2
Molecular Formula C2ClF2NaO2
Molecular Weight (g/mol) 152.457
MDL Number MFCD00064771
InChI Key MRTAVLDNYYEJHK-UHFFFAOYSA-M
Synonym sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium
PubChem CID 2734985
IUPAC Name sodium;2-chloro-2,2-difluoroacetate
SMILES C(=O)(C(F)(F)Cl)[O-].[Na+]

Specifications

Melting Point ∼195°C (decomposition)
Quantity 100 g
Beilstein 4162363
Solubility Information Soluble in dimethoxyethane,diglyme (0.60 g mL-1),triglyme,dimethyl formamide.
Formula Weight 152.46
Percent Purity 97%
Chemical Name or Material Sodium chlorodifluoroacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'