missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, 99+%, for analysis, Thermo Scientific Chemicals

€ 44.00 - € 87.13

Chemical Identifiers

CAS 6132-02-1
Molecular Formula CNa2O3·10H2O
Molecular Weight (g/mol) 286.14
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10348110
SDS View Documents
Thermo Scientific Acros
213500010
1 kg Plastic bottle
€ 44.00
1kg
Please to purchase this item. Need a web account? Register with us today!
10400301
SDS missing translation for 'viewDocuments'
Thermo Scientific Acros
213500025
2.5 kg Plastic bottle
€ 87.13
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Beschrijving

Beschrijving

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specificatie
Show More
Video's
Veiligheid en behandeling
Show More
SDS
Documentatie

Documentatie

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium carbonate decahydrate, 99+%, for analysis, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.