Learn More
Sodium 4-pentynoate, MedChemExpress
MedChemExpress Sodium 4-pentynoate is a alkynylacetate analogue, can be metabolically incorporated onto cellular proteins through biosynthetic mechanisms for profiling of acetylated proteins in diverse cell types.
Brand: Medchem Express HY-15286
Description
Sodium 4-pentynoate is a alkynylacetate analogue, can be metabolically incorporated onto cellular proteins through biosynthetic mechanisms for profiling of acetylated proteins in diverse cell types.Specifications
Sodium 4-pentynoate | |
101917-30-0 | |
100 mg | |
C5H5NaO2 | |
C#CCCC(O[Na])=O | |
120.08 | |
Research |
Solid | |
White | |
Research | |
H2O : 100 mg/mL (832.78 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (138.82 mM; Need ultrasonic) | |
120.08 | |
98.0% |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only