missing translation for 'onlineSavingsMsg'
Learn More
Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%, Thermo Scientific Chemicals Product Code.: 11489856

Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%, Thermo Scientific Chemicals

Product Code. 11489856
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11489856

Brand: Thermo Scientific Alfa Aesar B20332.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It can be used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as catalyst in pharmaceutical research.

Solubility
Solubility in water is almost transparent.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5838-34-6
Molecular Formula C20H14N4O4S
Molecular Weight (g/mol) 406.42
MDL Number MFCD00053627
InChI Key RFNALTSHRWGOHV-UHFFFAOYSA-N
Synonym sodium 1-undecanesulfonate, sodium undecane-1-sulfonate, 1-undecanesulfonic acid sodium salt, sodium undecylsulfonate, ipc-alks-11, acmc-1bn99, 1-undecanesulfonicacid, sodium salt 1:1
PubChem CID 23692564
IUPAC Name sodium;undecane-1-sulfonate
SMILES O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 3751377
Formula Weight 258.36
Percent Purity ≥98%
Assay (dry wt.), water <2%
Chemical Name or Material Sodium 1-undecanesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.