missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It can be used as catalyst in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used as catalyst in pharmaceutical research.
Solubility
Solubility in water is almost transparent.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It can be used as catalyst in pharmaceutical research.
Solubility
Solubility in water is almost transparent.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | >300°C |
Quantity | 25 g |
Beilstein | 3751377 |
Formula Weight | 258.36 |
Percent Purity | ≥98% |
Assay | (dry wt.), water <2% |
Chemical Name or Material | Sodium 1-undecanesulfonate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction