missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-propanesulfonate, 99%, Thermo Scientific Chemicals

Product Code. 11435836
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14533-63-2
Molecular Formula C3H7NaO3S
Molecular Weight (g/mol) 146.136
MDL Number MFCD00062546
InChI Key NPAWNPCNZAPTKA-UHFFFAOYSA-M
Synonym sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt
PubChem CID 4319363
IUPAC Name sodium;propane-1-sulfonate
SMILES CCCS(=O)(=O)[O-].[Na+]

Specifications

Quantity 1 g
Formula Weight 146.14
Percent Purity 99%
Chemical Name or Material Sodium 1-propanesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.