Learn More
Marienfeld Superior™ Uncoated Soda Lime Glass Microscope Slides
Product Code. 10056201
Click to view available options
Frosted:
Not Frosted
Silky Frosted Marking Area
Edges:
90°, Ground
Cut
Quantity:
50 Pieces in Cardboard Box
50/Pk.
Unit Size:
Pack of 2250
Pack of 2500
Product Code. 10056201
Brand: Marienfeld Superior™ 1000612
Only null left
Add to basket
Add to basket
Use Marienfeld Superior™ Microscope Slides for microscopic investigation and the preparation and archiving of specimens.
  • Well wettable microscope slides
  • Made of soda lime glass of third hydrolytic class
  • In compliance with DIN ISO 8037/1
  • Pre-cleaned, ready for use, autoclavable
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark, recommended best-before-date and batch number for comprehensive information and traceability
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
No. per Case 2500
No. per Pack 50
Edges 90°, Ground
Frosted Silky Frosted Marking Area
Material Soda Lime Glass Type III, Hydrolytic Class
Precleaned Yes
Thickness (Metric) 1 mm
Type Microscope Slide
Certifications/Compliance CE Mark, DIN ISO 8037/1
Corner Style 90°
Quantity 50 Pieces in Cardboard Box
Dimensions (L x W) 76 x 26 mm