Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Tube
Click to view available options
Length (Metric):
40 mm
50 mm
55 mm
70 mm
120 mm
125 mm
150 mm
Diameter (Metric):
8 mm
9.5 mm
10.5 mm
11 mm
12 mm
16 mm
Unit Size:
Pack of 200
Pack of 250
Pack of 500
Description
- Rimmed top
- Unlableled
- Clear soda lime glass
- Uniform bottom
- Disposable
Specifications
Specifications
| Design Type | Round bottom |
| Length (Metric) | 50 mm |
| Packaging | 200Pack |
| Diameter (Metric) | 9.5 mm |
| Product Type | Test Tube |