Learn More
Fisherbrand™ Soda Lime Glass Rimless Test Tube
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
75 mm
100 mm
150 mm
Diameter (Metric):
10 mm
12 mm
16 mm
18 mm
Unit Size:
Pack of 100
Description
- Soda Lime Glass
- Rimless
- Disposable
Specifications
Specifications
| Certifications/Compliance | ISO 4142 |
| Length (Metric) | 100 mm |
| Material | Soda Lime Glass |
| Quantity | 100 Pack |
| Rimmed | No |
| Diameter (Metric) | 12 mm |