Learn More
Thermo Scientific™ Snap-Cap Centrifuge Tubes, Plain
Shop All MBP Molecular BioProducts Products
Click to view available options
Color:
Natural
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Description
Thermo Scientific™ Plain Snap-Cap Centrifuge Tubes include a flat cap, and have no frosting or graduations.
- With flat cap
- No frosting or graduations
- Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
Specifications
Specifications
| Cap Style | Flat |
| Certifications/Compliance | Certified free of RNase, DNase, DNA and pyrogens |
| Description | Plain, Snap-Cap Centrifuge Tube |
| Capacity (Metric) | 0.6 mL |
| Color | Natural |
| Material | Polypropylene |
| Packaging | 10 bags of 100 |
| Sterility | Non-sterile |
| Graduated | No |
| Max. RCF | 25,000 x g |