Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin
Product Code. 30130372 Shop All Thermo Scientific Products
Click to view available options
Description:
Magnetic, SOLAμ Collection Plate, with Soluble Trypsin
Magnetic, with Soluble Trypsin
Magnetic, without Soluble Trypsin
Unit Size:
1 set
Product Code. 30130372
Brand: Thermo Scientific™ 60119103
Perform simple and easy streptavidin immunocapture for protein sample cleanup prior to protein digestion or analysis.
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin. These kits were designed to remove the challenges often associated with the process of protein cleanup by providing a simple and easy capture workflow.
The kit contains beads with an attached streptavidin moiety enabling a strong streptavidin-biotin bond with the appropriate biotinylated molecule with specific affinity for the target biomolecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
TRUSTED_SUSTAINABILITY
Specifications
Description Magnetic, SOLAμ Collection Plate, with Soluble Trypsin
Capture Streptavidin
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format Bulk Magnetic
For Use With (Application) Immunocapture and Protein Digest Sample Preparation
Product Line SMART Digest
Type Immunocapture + Protein Digestion (separate)
Includes Bulk Magnetic SMART Digest Resin, SMART Digest Soluble Trypsin, Collection Plate, SOLAμ SPE Plate
Product Content Correction
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato