Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G
Product Code. p-9078060 Shop All Thermo Scientific Products
Click to view available options
Description:
Magnetic, Soluble
Magnetic, Standalone
Magnetic, with SOLAμ Collection Plate, Soluble
Non-Magnetic, Soluble
Non-Magnetic, with SOLAμ Collection Plate, Soluble
Unit Size:
Each
Product Code. 15830182
Brand: Thermo Scientific™ 60115101
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
TRUSTED_SUSTAINABILITY
Specifications
Description Non-Magnetic, Soluble
Capture Protein G
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format Bulk Non-magnetic
For Use With (Application) Immunocapture and Protein Digest Sample Preparation
Product Line SMART Digest
Type SMART Digest Bulk Fractionation Kit
Includes Bulk Non-magnetic SMART Digest Soluble Resin