Learn More
Horst™ Slim Heating Tape with High Specific Heat Output
Double-laid heating conductors in a slim, insulated tape.
Brand: Horst™ 25202
This item is not returnable.
View return policy
Description
- Slim design.
- Electrically insulated with PTFE.
- Metallic protective braid of nickel-plated copper.
Specifications
Bending radius: 10 mm, Thickness: 4 mm | |
230 V | |
1 m | |
4 mm |
1.0 m | |
PTFE, Nickel-plated Copper | |
50 W | |
250°C (Nominal) |