Learn More
Horst™ Slim Heating Tape with High Specific Heat Output
Double-laid heating conductors in a slim, insulated tape.
Brand:  Horst™ 25203
 View more versions of this product
Product Code. 10458641
  • € 151.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Description
  • Slim design.
  • Electrically insulated with PTFE.
  • Metallic protective braid of nickel-plated copper.
Specifications
Specifications
Bending radius: 10 mm, Thickness: 4 mm
230 V
1 m
4 mm
2.0 m
PTFE, Nickel-plated Copper
100 W
250°C (Nominal)