Learn More
Epredia™ SlideMate™ AS Slide Printer
Reduce specimen identification errors while enhancing lean workflow with the Epredia™ SlideMate™ AS On-Demand Slide Printer.
Brand: Epredia™ A83900005
This item is not returnable.
View return policy
Description
The SlideMate AS is the next generation Epredia Slide Printer delivering a smaller footprint than previous models, making it even easier to fit next to the microtome. The SlideMate AS promotes single piece workflow though unique features designed specifically for on-demand printing.- Fully integrated color touch-screen and simple software are easy to use
- Build-in barcode scanner reads cassette information and imports the data to the slide label
- Connects easily to other computers or Laboratory Information Systems
- Prints directly onto the slide, eliminating the need for hand writing or labels
Specifications
Slide Printer AS On-Demand Sys | |
SlideMate AS on-demand printer with slide delivery system, color touch-screen, built-in scanner, two USB ports, one ethernet port, power cord and advanced software enabling label creation, special panel printing and import from LIS |
Product Content Correction
Product Title
Epredia™ SlideMate™ AS Slide Printer >