Learn More
Ibidi μ-Slide VI 0.5 Glass Bottom
μ-Slide VI 0.5 Glass Bottom is a 6 channel slide for parallel flow assays and immunofluorescence.
Brand: Ibidi 80607
Description
- A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
- bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
- sterilized
Specifications
Microscope Slide | |
15 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.