Learn More
Ibidi μ-Slide VI 0.4 ibiTreat, Bulk Pack
μ-Slide VI 0.4 ibiTreat, Bulk Pack is a 6 channel slide for parallel flow assays and immunofluorescence.
Brand: Ibidi 80606-90
This item is not returnable.
View return policy
Description
- A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Microscope Slide | |
90 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibidi μ-Slide VI 0.4 ibiTreat, Bulk Pack