Learn More
Kartell™ Slide Round Stirring Bar
Product Code. 10152231
Click to view available options
Length (Metric):
20, 57 mm
24, 76 mm
Unit Size:
Pack of 10
Product Code. 10152231
Brand: Kartell™ 0078700
Only null left
Add to basket
Add to basket
The pivot ring reduces the contact surface and fosters optimal stir bar positioning
  • Angled surfaces can provide lots of turbulence even at slow speeds
  •  For containers with irregular bottom - designed to maintain the optimum position in vessels with curved or uneven bottoms
  • The ring helps to reduce vibrations and wear, and reduce the effects of friction from contact with the vessel
  • Suitable for foodstuff
  • Specifications
    Coating PTFE
    Diameter (Metric) 6,19 mm
    Length (Metric) 24, 76 mm
    Material Magnet, PTFE
    Quantity 10