Learn More
Invitrogen™ SIRT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA116956
This item is not returnable.
View return policy
Description
PA1-16956 is specific for the ~39 kDa SIRT6 protein. A higher molecular weight band at ~59 kDa may be seen with lower dilutions such as 2 μg/mL.
Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
Specifications
| SIRT6 | |
| Polyclonal | |
| Unconjugated | |
| SIRT6 | |
| 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51548 | |
| 4°C, do not freeze | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Phosphate, tris citrate with 0.1% sodium azide; pH 7-8 | |
| Q8N6T7 | |
| SIRT6 | |
| A synthetic peptide made to an internal region of the human SIRT6 protein (within residues 250-350). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |