Learn More
Honeywell™ Single-Use Molded Facemask, FFP1 NRD with Valve
Click to view available options
:
Size: Medium
Description
- Rigid shell: shape maintained even in moistened and heavy loading environments
- Shielded exhalation valve for optimum safety
- Preformed and colored nosebridge to save time to perfectly adjust the mask and to instantly identify its protection level
- Large internal volume to easily communicate and breathe
- Twin elastic straps, ultra flexible and strong, stapled outside the filtering area to avoid any risk of leakages
- Lightweight (15 g) for better wearer acceptance
- Free of latex, silicone, and PVC
Specifications
Specifications
| Design Type | Cup/Molded |
| Size | Medium |
| With Exhalation Valve | Yes |
| For Use With (Application) | Building, construction, iron/steel, textile industry |
| Includes | Double elastics |
| Latex Free | Yes |
| Weight (Metric) | 15 g |