Learn More
Fisherbrand™ Single-Channel Pipette
Aspirates and dispenses viscous liquids accurately and reliably. Fisherbrand™ Single-Channel Pipette include a lock counter to prevent accidental volume changes. Lightweight, ergonomic design minimizes repetitive strain injuries.
Brand:  Fisherbrand™ FI8-105-01-9
 View more versions of this product
Product Code. 10544073
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Accurate and reliable pipetting
  • Reverse pipetting option for accurate and precise aspirating and dispensing of viscous liquids
  • Innovative soft grip prevents transfer of heat to internal components, ensuring accurate pipette operation
  • Lock counter mechanism of variable volume pipettes prevents volume changes while pipetting
  • Comfortable and effortless pipetting - light weight / ergonomic design for easy operation prevents hand stress. Ergonomic low plunger force minimizes RSI (repetitive strain injury)
  • Pipette is fully autoclavable for sterile handling
  • Easy and quick maintenance and adjustment with the included disassembly and calibration tool
  • No loss of sample - blow out system allows last drop dispensing
  • Meets following standards: DIN EN ISO 8655-1 | DIN EN ISO 8655-2 | DIN EN ISO 8655-6 | EU Directive 98/79/EG
Specifications
Specifications
Autoclavable
Sterile
50 to 200 μL
1.0μL
2 Years
1
Manual Pipette
Lock counter mechanism, disassembly and calibration tool, sample bag with tips, grease
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers