Learn More
Silver Sulfate Solution, Chem Lab™ Product Code.: 11909252
compliance-icons
Silver Sulfate Solution, Chem Lab™
Product Code. 11909252
2.5 L
Click to view available options
Quantity:
1 L
2.5 L
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11909252
Brand: Chem Lab CL02.2603.2500
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Specifications
Color Colorless
Boiling Point 330°C
Melting Point -5°C
Composition 10 g Ag2SO4 / l H2SO4 95-97%
Density 1.84g/mL
Solubility Information Water Soluble
Odor Odorless
pH <1
Quantity 2.5 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Silver sulphate 10 g/l solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Silver sulphate, Sulphuric acid
Product Content Correction
Product Title
Silver Sulfate Solution, Chem Lab™ >