missing translation for 'onlineSavingsMsg'
Learn More
Silver Sulfate, puriss. p.a. ACS Reagent, Honeywell Fluka™ Product Code.: 15633470

Silver Sulfate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Product Code. 15633470 Shop All Honeywell Fluka Products
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15633470

Brand: Honeywell-Fluka 31494100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number MFCD00003407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Quantity 100 g
Linear Formula Ag2SO4
UN Number 3077
Formula Weight 311.8g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Chemical Name or Material Silver Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver Sulfate, puriss. p.a. ACS Reagent, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.