Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Shop All Thermo Scientific ProductsDescription
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Repair broken columns
Specifications
Specifications
| Type | Replacement Column Connector Nuts |
| Description | 290 MN211 |
| Diameter (Metric) Inner | 0.1 to 0.53 mm |
| For Use With (Equipment) | Fused Silica Capillary Column |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.