Learn More
Thermo Scientific™ SilTite™ Ferrules
Product Code. 11560964 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Pack of 10
Product Code. 11560964
Brand: Thermo Scientific™ 290MF231
Only null left
Add to basket
Add to basket
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Description For 0.53 mm I.D. Capillary; Pk. of 10
Diameter (Metric) 0.53 mm
For Use With (Equipment) Fused Silica Capillary GC Columns
Material SilTite Metal
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.