missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Silkam™ Needle-Thread Combination

Product Code. p-7160140
Click to view available options
Length (Metric):
0.7 mm
Needle Point Number:
DS 16
Shape:
3/8 Circle Reverse Cutting Needle
Size:
USP metric 6/0
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15317425

Brand: B Braun™ C0762113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A natural, non-absorbable braided and coated suture made of natural silk filaments.

  • Indicated for applications where it is surgical practice to employ non-absorbable material
  • Excellent handling properties
  • Good knot security
  • Indications include general surgery, oral surgery, neurosurgery, skin closure and ligatures

Specifications

Material Silk
Product Type Needle-thread Suture Combo
Length (Metric) 0.7 mm
Size USP metric 6/0
For Use With (Application) Surgical
Color Black
Shape 3/8 Circle Reverse Cutting Needle
Needle Point Number DS 16
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.