Learn More
Hirschmann™ SILICOSEN™ Silicone Caps
Click to view available options
Diameter (Metric):
20 to 30 mm
30 to 40 mm
40 to 55 mm
Unit Size:
Pack of 10
Description
- Uniform pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
Specifications
| Diameter (Metric) | 30 to 40 mm |
| For Use With (Application) | For The Preparation And Sterilization Of Culture Media |
| Length (Metric) | 28 mm |
| Quantity | 10/Pk. |
| Color | Pink |
| Material | Silicone |
| Product Type | Silicone Stopper |