Learn More
Deutsch Neumann™ Silicone Tubing
Product Code. 16204001
Click to view available options
Diameter (Metric) Inner:
1 mm
2 mm
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
9 mm
10 mm
12 mm
Diameter (Metric) Outer:
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
13 mm
14 mm
16 mm
Unit Size:
1m
25m
5m
Product Code. 16204001
Brand: Deutsch Neumann™ 3100508
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Appropriate for use with peristaltic pumps. Deutsch Neumann™ Silicone Tubing is transparent, certified food grade and suitable for general laboratory usage.
  • Temperature range of -60°C to +200°C (short periods up to +260°C)
  • Meets following standards: BFR XV, FDA CFR 21 § 177.2600, EP - Europ. Pharmacopoeia
  • Test for biocompatibility according to ISO 10993-1
  • Durometer 60 Shore A
Specifications
Certifications/Compliance BFR XV , FDA 177.2600 , DAB 10 , Ph Eur.2.Edition
Diameter (Metric) Inner 5 mm
Material Silicone
Diameter (Metric) Outer 8 mm
Overall Length (Metric) 25 m
Thickness (Metric) 1.5 mm
For Use With Peristaltic Pumps
Min. Operating Temperature (Metric) -60°C
Packaging Quantity 1 Roll
Color Transparent
Hardness 60 (Shore A, ±5)
Max. Operating Temperature (Metric) 200°C