Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
Click to view available options
Diameter (Metric) Bottom:
3.5 mm
10.5 mm
14 mm
21 mm
29 mm
47 mm
Unit Size:
Each
Pack of 100
Description
- Acc. to DIN 12 871
- Durometer 50 Shore A
- Available in a wide range of sizes
Specifications
Specifications
| Diameter (Metric) Bottom | 47 mm |
| Diameter (Metric) Top | 55 mm |
| Material | Silicone |
| Product Type | Poly Stopper |