Learn More
Saint-Gobain Silicone Iso-Versinic™ Stopper
Saint Gobain Performance Plastics™ Silicone Iso-Versinic™ Stoppers have high chemical resistance especially to hydrocarbons, oils, strong acids and aromatic and chlorinated solvents.
Brand:  Saint-Gobain 263070-02
 View more versions of this product
Product Code. 11332064
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Withstands temperatures between -20° and 200°C (possible peaks to 200°C)
  • High chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For collar and tube
60 mm Bottom Diameter
60 mm
Silicone
50 mm
–20 to 200°C
70 mm
ISO-Versinic™ Stoppers