Learn More
Deutsch Neumann™ Vacuum Quality Silicone Tubing
Provides flexibility, tear strength and resilience for vacuum applications. Deutsch Neumann™ Vacuum Quality Silicone Tubing is transparent, certified food grade and suitable for general laboratory usage.
Brand:  Deutsch Neumann™ 3110612
Product Code. 10615801
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Temperature range of -60°C to +200°C (short periods up to +260°C)
  • Meets following standards: BFR XV, FDA CFR 21 § 177.2600, EP - Europ. Pharmacopoeia
  • Test for biocompatibility according to ISO 10993-1
  • Durometer 60 Shore A
Specifications
Specifications
6 mm
2500 cm
Transparent
12 mm
3 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Deutsch Neumann™ Vacuum Quality Silicone Tubing > Color: Transparent: Length: 25m