Learn More
Thermo Scientific Chemicals Silica gel 60, 0.075-0.2mm (70-200 mesh) Product Code.: 11315497
Thermo Scientific Chemicals Silica gel 60, 0.075-0.2mm (70-200 mesh)
Product Code. 11315497
-70/200 Mesh, 50 g, 470 to 570 m2/g
Click to view available options
Particle Size:
-70/200 Mesh
70/150 Mesh
Quantity:
50 g
250 g
1 kg
5 kg
Surface Area (Metric):
470 to 570 m2/g
500 to 600 m2/g
Product Code. 11315497
Brand: Thermo Scientific Chemicals 042723.18
Request Bulk or Custom Format
Classic column chromatography with no applied pressure (gravity pulled)Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size -70/200 Mesh
Pore Size 60 Å
Packaging Bottle
pH 6.5 to 7.5
Quantity 50 g
Surface Area (Metric) 470 to 570 m2/g
Type Silica Gel
Material Silica
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX15K011 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number042723.A7,042723.18,042723.A1,042723.30
Lot NumberR21K023 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number042723.A7,042723.18,042723.A1,042723.30
Lot NumberP14K028 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number042723.A7,042723.18,042723.A1,042723.30
Lot NumberZ07I009 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog Number042723.A7,042723.18,042723.A1,042723.30
Lot NumberU16I003 Certificate TypeCertificate of Analysis Date23/08/2022 Catalog Number042723.A7,042723.18,042723.A1,042723.30
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Silica gel 60, 0.075-0.2mm (70-200 mesh) >