Learn More
SymLine™ Signal Boxes
Product Code. 17853520
Click to view available options
Length (Metric):
3 m
55 mm
86 mm
Product Type:
Signalbox T1
Signalbox T5
Plug Type:
EU-style
UK-style
US-style
Includes:
1 Sensor Input and 1 Potential-free Output
5 Sensor Inputs and 6 Potential-free Outputs
Alarm at Empty State
Alarm at Full State
T5 with 5 Disc Fill Level Sensors
Unit Size:
Each
Product Code. 17853520
Brand: SymLine™ 108159
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Monitor two filling levels at once. Ideal for integration into lab furniture with SymLine™ Built-in Signal Box.
  • Clever way to change disposal container without interrupting the inflow
  • LEDs and acoustic buzzer warn you before filling level of collection containers reaches critical height
  • When container is changed, alert can be muted via new touchpad
Specifications
Product Type Signalbox T1
For Use With (Equipment) SCAT Sensors and Safety Waste Caps
Length (Metric) 3 m
Plug Type US-style
Size 180 x 105 x 55 mm
Voltage 110 V
For Use With (Application) Monitoring Filling Levels
Includes Alarm at Empty State